siRNA Details
| Gene Symbol: Olfr1446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146704.1 | NP_666915.1 | 1 | 252 | ||
| GenBank mRNA | BC119061.1 | - | 1 | 335 | 8733810 | |
| BC137821.1 | - | 1 | 335 | 8860840 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1446| Gene Symbol: Olfr1446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146704.1 | NP_666915.1 | 1 | 252 | ||
| GenBank mRNA | BC119061.1 | - | 1 | 335 | 8733810 | |
| BC137821.1 | - | 1 | 335 | 8860840 | ||