siRNA Details
| Gene Symbol: Olfr1271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146793.1 | NP_667004.1 | 1 | 571 | ||
| GenBank mRNA | BC106779.1 | - | 1 | 571 | 40040295 | |
| BC106780.1 | - | 1 | 571 | 40040533 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1271| Gene Symbol: Olfr1271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146793.1 | NP_667004.1 | 1 | 571 | ||
| GenBank mRNA | BC106779.1 | - | 1 | 571 | 40040295 | |
| BC106780.1 | - | 1 | 571 | 40040533 | ||