siRNA Details
| Gene Symbol: Olfr545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146840.1 | NP_667051.1 | 1 | 320 | ||
| GenBank mRNA | BC106987.1 | - | 1 | 320 | 40043277 | |
| BC128265.1 | - | 1 | 320 | 40043266 | ||
| BC128266.1 | - | 1 | 320 | 40043268 | ||

Olfr545
Genomic Map