siRNA Details
| Gene Symbol: Hgsnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029884.1 | NP_084160.1 | 18 | 2263 | ||
| GenBank mRNA | AK020410.1 | - | 3 | 590 | ||
| AK035264.1 | - | 18 | 2196 | |||
| AK149883.1 | - | 18 | 2267 | |||
| AK152926.1 | - | 18 | 2228 | |||
| AK159649.1 | - | 18 | 2192 | |||
| AK160068.1 | - | 18 | 2186 | |||
| BC019450.1 | - | 2 | 494 | |||
| BC024084.1 | - | 18 | 2170 | 5101064 | ||

Hgsnat
Genomic Map