siRNA Details
Gene Symbol: Vmn1r33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134169.1 | NP_598930.1 | 1 | 757 | ||
GenBank mRNA | BC138428.1 | - | 1 | 806 | 8861448 | |
BC138429.1 | - | 1 | 806 | 8861449 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134169.1 | NP_598930.1 | 1 | 757 | ||
GenBank mRNA | BC138428.1 | - | 1 | 806 | 8861448 | |
BC138429.1 | - | 1 | 806 | 8861449 |