siRNA Details
Gene Symbol: Vmn1r231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134196.1 | NP_598957.1 | 1 | 218 | ||
GenBank mRNA | AF394947.1 | - | 287 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134196.1 | NP_598957.1 | 1 | 218 | ||
GenBank mRNA | AF394947.1 | - | 287 |