siRNA Details
| Gene Symbol: Diras1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145217.2 | NP_660252.1 | 2 | 531 | ||
| GenBank mRNA | AK081336.1 | - | 2 | 531 | ||
| AY059640.1 | - | 1 | 437 | |||
| BC057556.1 | - | 2 | 531 | 6810812 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Diras1| Gene Symbol: Diras1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145217.2 | NP_660252.1 | 2 | 531 | ||
| GenBank mRNA | AK081336.1 | - | 2 | 531 | ||
| AY059640.1 | - | 1 | 437 | |||
| BC057556.1 | - | 2 | 531 | 6810812 | ||