siRNA Details
| Gene Symbol: ASIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001672.2 | NP_001663.2 | 2, 3 | 243 | ||
| XM_005260412.3 | XP_005260469.1 | 3, 4 | 2299 | |||
| XM_011528820.2 | XP_011527122.1 | 4, 5 | 422 | |||
| XM_011528821.1 | XP_011527123.1 | 4, 5 | 380 | |||
| GenBank mRNA | BC104238.1 | - | 2, 3 | 235 | 40033052 | |
| BC104239.2 | - | 2, 3 | 235 | 40033056 | ||

ASIP
Genomic Map