siRNA Details
| Gene Symbol: Olfr599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146731.1 | NP_666942.1 | 1 | 216 | ||
| GenBank mRNA | BC141003.1 | - | 1 | 260 | 9055615 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr599| Gene Symbol: Olfr599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146731.1 | NP_666942.1 | 1 | 216 | ||
| GenBank mRNA | BC141003.1 | - | 1 | 260 | 9055615 | |