siRNA Details
| Gene Symbol: Olfr921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146782.2 | NP_666993.2 | 3 | 924 | ||
| GenBank mRNA | BC131918.1 | - | 1 | 585 | 40130195 | |
| BC131920.1 | - | 1 | 585 | 40130197 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr921| Gene Symbol: Olfr921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146782.2 | NP_666993.2 | 3 | 924 | ||
| GenBank mRNA | BC131918.1 | - | 1 | 585 | 40130195 | |
| BC131920.1 | - | 1 | 585 | 40130197 | ||