siRNA Details
| Gene Symbol: Olfr1097 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC104098.1 | - | 1 | 465 | 40043068 | |
| BC104109.1 | - | 1 | 463 | |||
| BC106801.1 | - | 1 | 465 | 40042504 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1097| Gene Symbol: Olfr1097 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC104098.1 | - | 1 | 465 | 40043068 | |
| BC104109.1 | - | 1 | 463 | |||
| BC106801.1 | - | 1 | 465 | 40042504 | ||