siRNA Details
| Gene Symbol: Olfr170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146957.1 | NP_667168.1 | 1 | 596 | ||
| GenBank mRNA | BC132585.1 | - | 1 | 633 | 40130862 | |
| BC132587.1 | - | 1 | 633 | 40130864 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr170| Gene Symbol: Olfr170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146957.1 | NP_667168.1 | 1 | 596 | ||
| GenBank mRNA | BC132585.1 | - | 1 | 633 | 40130862 | |
| BC132587.1 | - | 1 | 633 | 40130864 | ||