siRNA Details
| Gene Symbol: Olfr610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147081.2 | NP_667292.2 | 1 | 77 | ||
| GenBank mRNA | BC113199.1 | - | 1 | 76 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr610| Gene Symbol: Olfr610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147081.2 | NP_667292.2 | 1 | 77 | ||
| GenBank mRNA | BC113199.1 | - | 1 | 76 | ||