siRNA Details
| Gene Symbol: Olfr978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147105.2 | NP_667316.2 | 1 | 406 | ||
| GenBank mRNA | BC114537.1 | - | 1 | 405 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr978| Gene Symbol: Olfr978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147105.2 | NP_667316.2 | 1 | 406 | ||
| GenBank mRNA | BC114537.1 | - | 1 | 405 | ||