siRNA Details
| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170755.2 | NP_739561.2 | 2 | 393 | ||
| GenBank mRNA | AK034667.1 | - | 2 | 394 | ||
| AK152268.1 | - | 2 | 108 | |||
| AK158940.1 | - | 2 | 163 | |||
| AK160400.1 | - | 2 | 347 | |||
| AK165957.1 | - | 2 | 372 | |||
| AK172093.1 | - | 2 | 390 | |||
| BC070406.1 | - | 2 | 385 | 5707319 | ||
| BY261213.1 | - | 2 | 171 | |||

Fam134a
Genomic Map