siRNA Details
| Gene Symbol: Fam160a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172682.3 | NP_766270.2 | 5 | 1174 | ||
| GenBank mRNA | AK036881.1 | - | 2 | 160 | ||
| BC043298.1 | - | 2 | 126 | |||
| BC094511.1 | - | 3 | 385 | |||
| DV651332.1 | - | 3 | 565 | |||

Fam160a1
Genomic Map