siRNA Details
| Gene Symbol: Bricd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175682.3 | NP_783613.1 | 2, 3 | 516 | ||
| GenBank mRNA | AK079437.1 | - | 2, 3 | 517 | ||
| BC096398.1 | - | 2, 3 | 277 | |||
| BC119389.1 | - | 3, 4 | 302 | 8734138 | ||
| BC120670.1 | - | 3, 4 | 302 | 40129593 | ||
| BC145010.1 | - | 3, 4 | 302 | 9053558 | ||

Bricd5
Genomic Map