siRNA Details
| Gene Symbol: Olfr1226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146967.1 | NP_667178.1 | 1 | 911 | ||
| GenBank mRNA | BC106781.1 | - | 1 | 911 | 40040933 | |
| BC106782.1 | - | 1 | 911 | 40040934 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1226| Gene Symbol: Olfr1226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146967.1 | NP_667178.1 | 1 | 911 | ||
| GenBank mRNA | BC106781.1 | - | 1 | 911 | 40040933 | |
| BC106782.1 | - | 1 | 911 | 40040934 | ||