siRNA Details
Gene Symbol: Olfr173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147000.2 | NP_667211.2 | 1 | 548 | ||
GenBank mRNA | BC120572.1 | - | 1 | 548 | 40129495 | |
BC137837.1 | - | 1 | 548 | 8860856 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147000.2 | NP_667211.2 | 1 | 548 | ||
GenBank mRNA | BC120572.1 | - | 1 | 548 | 40129495 | |
BC137837.1 | - | 1 | 548 | 8860856 |