siRNA Details
| Gene Symbol: Olfr166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147068.1 | NP_667279.1 | 1 | 913 | ||
| GenBank mRNA | BC116856.1 | - | 1 | 960 | 40126175 | |
| BC119189.1 | - | 1 | 960 | 8733938 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr166| Gene Symbol: Olfr166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147068.1 | NP_667279.1 | 1 | 913 | ||
| GenBank mRNA | BC116856.1 | - | 1 | 960 | 40126175 | |
| BC119189.1 | - | 1 | 960 | 8733938 | ||