siRNA Details
| Gene Symbol: Olfr641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147072.1 | NP_667283.1 | 1 | 256 | ||
| GenBank mRNA | BC127981.1 | - | 1 | 260 | 40135228 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr641| Gene Symbol: Olfr641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147072.1 | NP_667283.1 | 1 | 256 | ||
| GenBank mRNA | BC127981.1 | - | 1 | 260 | 40135228 | |