siRNA Details
Gene Symbol: Olfr610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147081.2 | NP_667292.2 | 1 | 237 | ||
GenBank mRNA | BC113199.1 | - | 1 | 236 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147081.2 | NP_667292.2 | 1 | 237 | ||
GenBank mRNA | BC113199.1 | - | 1 | 236 |