siRNA Details
Gene Symbol: Defb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153108.4 | NP_694748.3 | 2 | 165 | ||
GenBank mRNA | AJ300674.1 | - | 2 | 165 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Defb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153108.4 | NP_694748.3 | 2 | 165 | ||
GenBank mRNA | AJ300674.1 | - | 2 | 165 |