siRNA Details
| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170755.2 | NP_739561.2 | 3, 4 | 473 | ||
| GenBank mRNA | AK034667.1 | - | 3, 4 | 474 | ||
| AK152268.1 | - | 3, 4 | 188 | |||
| AK158940.1 | - | 3, 4 | 243 | |||
| AK160400.1 | - | 3, 4 | 427 | |||
| AK165957.1 | - | 3, 4 | 452 | |||
| AK172093.1 | - | 3, 4 | 470 | |||
| BC070406.1 | - | 3, 4 | 465 | 5707319 | ||
| BY261213.1 | - | 3, 4 | 251 | |||

Fam134a
Genomic Map