siRNA Details
| Gene Symbol: Svs3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173377.2 | NP_775553.1 | 2, 3 | 871 | ||
| GenBank mRNA | AK035235.1 | - | 2, 3 | 869 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Svs3b| Gene Symbol: Svs3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173377.2 | NP_775553.1 | 2, 3 | 871 | ||
| GenBank mRNA | AK035235.1 | - | 2, 3 | 869 | ||