siRNA Details
| Gene Symbol: Nrn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301174.1 | NP_001288103.1 | 3 | 334 | ||
| NM_175024.4 | NP_778189.1 | 3 | 337 | |||
| GenBank mRNA | AK080711.1 | - | 3 | 332 | ||
| AK090312.1 | - | 3 | 335 | |||
| BC100315.1 | - | 3 | 337 | 30931461 | ||
| CK030777.1 | - | 3 | 318 | |||
| DQ176852.1 | - | 3 | 324 | |||

Nrn1l
Genomic Map