siRNA Details
| Gene Symbol: 1500011K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005218.1 | - | 1 | 312 | ||
| AK008527.1 | - | 1 | 307 | |||
| AK027942.1 | - | 1 | 307 | |||
| AK165423.1 | - | 1 | 292 | |||
| BC048556.1 | - | 1 | 301 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1500011K16Rik| Gene Symbol: 1500011K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005218.1 | - | 1 | 312 | ||
| AK008527.1 | - | 1 | 307 | |||
| AK027942.1 | - | 1 | 307 | |||
| AK165423.1 | - | 1 | 292 | |||
| BC048556.1 | - | 1 | 301 | |||