siRNA Details
| Gene Symbol: Ttbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162864.1 | NP_001156336.1 | 15 | 6783 | ||
| GenBank mRNA | AK049158.1 | - | 1 | 1745 | ||
| AK173279.1 | - | 2 | 3747 | |||
| BC059249.1 | - | 2 | 3987 | |||
| BC094327.1 | - | 1 | 2068 | |||

Ttbk1
Genomic Map