siRNA Details
| Gene Symbol: Tnfrsf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178931.2 | NP_849262.1 | 3 | 242 | ||
| GenBank mRNA | AF515707.1 | - | 3 | 211 | ||
| AY264405.1 | - | 3 | 242 | |||
| BC104054.1 | - | 3 | 242 | 40040309 | ||
| BC104055.1 | - | 3 | 242 | 40040311 | ||

Tnfrsf14
Genomic Map