siRNA Details
| Gene Symbol: Opn1mw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008106.2 | NP_032132.1 | 1, 2 | 138 | ||
| GenBank mRNA | AF011389.1 | - | 1, 2 | 124 | ||
| AF190672.1 | - | 1, 2 | 140 | |||
| AK149287.1 | - | 1, 2 | 139 | |||
| BC014826.1 | - | 1, 2 | 118 | 4507311 | ||
More Information
Set Membership: |
|

Opn1mw
Genomic Map