siRNA Details
| Gene Symbol: BPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001725.2 | NP_001716.2 | 2 | 244 | ||
| GenBank mRNA | AF322588.1 | - | 2 | 170 | ||
| AK296568.1 | - | 2 | 89 | |||
| AK315328.1 | - | 2 | 211 | |||
| AX713278.1 | - | 2 | 212 | |||
| AX746123.1 | - | 2 | 212 | |||
| AX752831.1 | - | 2 | 212 | |||
| BC040955.1 | - | 2 | 219 | 5218655 | ||
| BP306215.1 | - | 2 | 256 | |||
| DQ414688.1 | - | 2 | 182 | |||
| J04739.1 | - | 2 | 212 | |||

BPI
Genomic Map