siRNA Details
| Gene Symbol: Hoxc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175730.5 | NP_783857.1 | 1 | 499 | ||
| GenBank mRNA | AK078527.1 | - | 1 | 500 | ||
| BC115551.1 | - | 1 | 500 | 40101712 | ||
| BC115552.1 | - | 1 | 500 | 40101713 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108116.

Hoxc5
Genomic Map