siRNA Details
| Gene Symbol: Klk1b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010915.3 | NP_035045.2 | 93 | |||
| GenBank mRNA | BC034518.1 | - | 93 | 4911885 | ||
| M11434.1 | - | 89 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk1b4| Gene Symbol: Klk1b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010915.3 | NP_035045.2 | 93 | |||
| GenBank mRNA | BC034518.1 | - | 93 | 4911885 | ||
| M11434.1 | - | 89 | ||||