siRNA Details
| Gene Symbol: Sprr2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011474.3 | NP_035604.1 | 2 | 385 | ||
| GenBank mRNA | AJ005566.1 | - | 1 | 357 | ||
| AY158992.1 | - | 2 | 385 | |||
| BC099888.1 | - | 3 | 154 | |||
| BC099889.1 | - | 2 | 371 | |||
| BC099890.1 | - | 2 | 88 | |||
| BC127024.1 | - | 2 | 368 | |||

Sprr2h
Genomic Map