siRNA Details
| Gene Symbol: Ttn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011652.3 | NP_035782.3 | 310 | 99411 | ||
| NM_028004.2 | NP_082280.2 | 189 | 79668 | |||
| GenBank mRNA | AK029024.1 | - | 3 | 509 | ||
| BC025840.1 | - | 1 | 162 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttn| Gene Symbol: Ttn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011652.3 | NP_035782.3 | 310 | 99411 | ||
| NM_028004.2 | NP_082280.2 | 189 | 79668 | |||
| GenBank mRNA | AK029024.1 | - | 3 | 509 | ||
| BC025840.1 | - | 1 | 162 | |||