siRNA Details
| Gene Symbol: C5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317163.1 | NP_001304092.1 | 2 | 569 | ||
| NM_001317164.1 | NP_001304093.1 | 2 | 240 | |||
| NM_001735.2 | NP_001726.2 | 2 | 243 | |||
| XM_011518980.2 | XP_011517282.1 | 3 | 612 | |||
| XM_017015102.1 | XP_016870591.1 | 4 | 764 | |||
| XM_017015103.1 | XP_016870592.1 | 2 | 397 | |||
| XM_017015104.1 | XP_016870593.1 | 4 | 764 | |||
| GenBank mRNA | AB209031.1 | - | 2 | 569 | ||
| AK310774.1 | - | 2 | 240 | |||
| AK310780.1 | - | 2 | 242 | |||
| BC113738.1 | - | 2 | 238 | 8322790 | ||
| BC113740.1 | - | 2 | 238 | 8322792 | ||
| M57729.1 | - | 2 | 225 | |||

C5
Genomic Map