siRNA Details
| Gene Symbol: Olfr65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013616.4 | NP_038644.2 | 1 | 908 | ||
| GenBank mRNA | BC137766.1 | - | 1 | 908 | 8860785 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr65| Gene Symbol: Olfr65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013616.4 | NP_038644.2 | 1 | 908 | ||
| GenBank mRNA | BC137766.1 | - | 1 | 908 | 8860785 | |