siRNA Details
| Gene Symbol: Timm8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013897.2 | NP_038925.1 | 2 | 180 | ||
| GenBank mRNA | AF196314.1 | - | 169 | |||
| AK003382.1 | - | 2 | 139 | |||
| AK004190.1 | - | 169 | ||||
| AK012925.1 | - | 2 | 147 | |||
| AK018718.1 | - | 2 | 147 | |||
| AK019422.1 | - | 2 | 155 | |||
| AK169258.1 | - | 2 | 159 | |||
| BC029239.1 | - | 2 | 133 | 4237232 | ||
| BY100901.1 | - | 2 | 184 | |||

Timm8b
Genomic Map