siRNA Details
| Gene Symbol: Isg15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015783.3 | NP_056598.2 | 2 | 197 | ||
| GenBank mRNA | BC031424.1 | - | 2 | 164 | 3670747 | |
| BC083156.1 | - | 2 | 146 | 5338191 | ||
| BC109346.2 | - | 2 | 147 | 40055493 | ||
| CT010333.1 | - | 1 | 87 | |||
| X56602.1 | - | 2 | 198 | |||
| Gene Symbol: Gm9706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019325.1 | - | 1 | 771 | ||

Isg15
Genomic Map