siRNA Details
| Gene Symbol: Ptprcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016933.3 | NP_058629.2 | 2 | 158 | ||
| GenBank mRNA | AA265746.1 | - | 2 | 157 | ||
| AI323196.1 | - | 2 | 130 | |||
| AK088334.1 | - | 2 | 156 | |||
| AK165203.1 | - | 2 | 156 | |||
| BB840651.1 | - | 2 | 156 | |||
| BC013273.1 | - | 2 | 157 | 4208992 | ||
| BC100560.1 | - | 2 | 139 | 832557 | ||
| D10105.1 | - | 2 | 134 | |||
| U03856.1 | - | 1 | 151 | |||

Ptprcap
Genomic Map