siRNA Details
Gene Symbol: Msmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020597.3 | NP_065622.1 | 3 | 192 | ||
GenBank mRNA | U89840.1 | - | 2 | 160 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Msmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020597.3 | NP_065622.1 | 3 | 192 | ||
GenBank mRNA | U89840.1 | - | 2 | 160 |