siRNA Details
| Gene Symbol: Krtcap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025327.2 | NP_079603.1 | 3 | 259 | ||
| GenBank mRNA | AK002479.1 | - | 3 | 260 | ||
| AK003310.1 | - | 3 | 227 | |||
| BC031492.1 | - | 3 | 212 | 3485092 | ||
| BC043030.1 | - | 3 | 731 | 5693076 | ||

Krtcap2
Genomic Map