siRNA Details
| Gene Symbol: Ifitm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025378.2 | NP_079654.1 | 1 | 183 | ||
| GenBank mRNA | AK003407.1 | - | 1 | 184 | ||
| AK007916.1 | - | 1 | 176 | |||
| AK007919.1 | - | 1 | 172 | |||
| AK151890.1 | - | 1 | 149 | |||
| AY082484.1 | - | 1 | 152 | |||
| AY594690.1 | - | 1 | 199 | |||
| BC010291.1 | - | 1 | 135 | 3152819 | ||

Ifitm3
Genomic Map