siRNA Details
| Gene Symbol: Rgcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025427.2 | NP_079703.2 | 2, 3 | 380 | ||
| GenBank mRNA | AF276981.1 | - | 2, 3 | 392 | ||
| AK004451.1 | - | 2, 3 | 381 | |||
| AK004705.1 | - | 2, 3 | 376 | |||
| BC049580.1 | - | 2, 3 | 376 | 6741281 | ||
| BC050935.1 | - | 2, 3 | 369 | 6818687 | ||
| CT010403.1 | - | 2, 3 | 242 | |||

Rgcc
Genomic Map