siRNA Details
| Gene Symbol: Snrpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026506.2 | NP_080782.1 | 3 | 203 | ||
| GenBank mRNA | AK011129.1 | - | 3 | 202 | ||
| AK012802.1 | - | 3 | 192 | |||
| AK013235.1 | - | 3 | 204 | |||
| AK017852.1 | - | 3 | 552 | |||
| BC027499.1 | - | 3 | 176 | 1430048 | ||
| BC051470.1 | - | 3 | 248 | 1244608 | ||
| BC094411.1 | - | 3 | 190 | 1380524 | ||

Snrpg
Genomic Map