siRNA Details
| Gene Symbol: Cmtm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027294.2 | NP_081570.1 | 2 | 422 | ||
| GenBank mRNA | AK012262.1 | - | 2 | 423 | ||
| AY243513.1 | - | 2 | 457 | |||
| BC027554.1 | - | 2 | 344 | 1382818 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmtm8| Gene Symbol: Cmtm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027294.2 | NP_081570.1 | 2 | 422 | ||
| GenBank mRNA | AK012262.1 | - | 2 | 423 | ||
| AY243513.1 | - | 2 | 457 | |||
| BC027554.1 | - | 2 | 344 | 1382818 | ||