siRNA Details
| Gene Symbol: Mier2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027422.2 | NP_081698.2 | 14 | 2080 | ||
| GenBank mRNA | AK012576.1 | - | 2 | 928 | ||
| AK154672.1 | - | 14 | 2097 | |||
| AK173128.1 | - | 13 | 2235 | |||
| BC005748.1 | - | 1 | 488 | |||
| BC067013.1 | - | 14 | 2080 | 6810181 | ||
| BC080757.1 | - | 7 | 1331 | |||
| BC083321.1 | - | 5 | 1201 | |||

Mier2
Genomic Map