siRNA Details
Gene Symbol: 4632415L05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014580.1 | - | 1 | 157 | ||
AW121548.1 | - | 1 | 207 | |||
BC023403.1 | - | 1 | 179 | 5252327 | ||
BC037267.1 | - | 1 | 167 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4632415L05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014580.1 | - | 1 | 157 | ||
AW121548.1 | - | 1 | 207 | |||
BC023403.1 | - | 1 | 179 | 5252327 | ||
BC037267.1 | - | 1 | 167 |