siRNA Details
| Gene Symbol: Lce1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028625.2 | NP_082901.1 | 2 | 639 | ||
| GenBank mRNA | AK003412.1 | - | 2 | 640 | ||
| BC062142.1 | - | 2 | 616 | |||
| BC080817.1 | - | 2 | 616 | |||
| BC100540.1 | - | 2 | 626 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lce1a2| Gene Symbol: Lce1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028625.2 | NP_082901.1 | 2 | 639 | ||
| GenBank mRNA | AK003412.1 | - | 2 | 640 | ||
| BC062142.1 | - | 2 | 616 | |||
| BC080817.1 | - | 2 | 616 | |||
| BC100540.1 | - | 2 | 626 | |||