siRNA Details
| Gene Symbol: Fam98c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146023.1 | NP_001139495.1 | 7, 8 | 923 | ||
| GenBank mRNA | AK003496.1 | - | 6, 7 | 847 | ||
| AK080790.1 | - | 6, 7 | 1015 | |||
| AK138395.1 | - | 6, 7 | 1826 | |||
| AK166139.1 | - | 6, 7 | 1822 | |||
| AK171108.1 | - | 5, 6 | 1289 | |||
| BC023421.1 | - | 4, 5 | 541 | 5029298 | ||
| BC058765.1 | - | 6, 7 | 980 | |||

Fam98c
Genomic Map